Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nefiracetam 98.0+%, TCI America™
SDP

Catalog No. N09961G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09961G 1 g
N0996200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09961G Supplier TCI America Supplier No. N09961G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 77191-36-7
Molecular Formula C14H18N2O2
Molecular Weight (g/mol) 246.31
MDL Number MFCD00209882
InChI Key NGHTXZCKLWZPGK-UHFFFAOYSA-N
Synonym nefiracetam, n-2,6-dimethylphenyl-2-2-oxopyrrolidin-1-yl acetamide, translon, nefiracetam inn, dmppa, motiva, nefiracetamum inn-latin, n-2,6-dimethylphenyl-2-oxo-1-pyrrolidineacetamide, unii-1jk12gx30n, ccris 6729
PubChem CID 71157
IUPAC Name N-(2,6-dimethylphenyl)-2-(2-oxopyrrolidin-1-yl)acetamide
SMILES CC1=C(C(=CC=C1)C)NC(=O)CN2CCCC2=O

Specifications

Melting Point 151°C
Color White
Quantity 1 g
Formula Weight 246.31
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Nefiracetam
RTECSNumber UX9655650
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.