Learn More
Medchemexpress LLC Nefopam (hydrochloride) | 23327-57-3 | 99.9% | 100 MG
SDP
Catalog No. 5000411769
Encompass_Preferred
Nefopam hydrochloride is a centrally-acting, non-opioid, and non-steroidal analgesic drug. It is derived from benzoxazocine and is used for the relief of moderate to severe pain. It targets β-catenin protein levels in mesenchymal cells both in-vitro and in-vivo, inhibiting β-catenin mediated signaling during skin wound repair.
  • Centrally-acting analgesic
  • Non-opioid and non-steroidal properties
  • Aids in the relief of moderate to severe pain
  • Inhibits β-catenin mediated signaling
  • Derived from benzoxazocine
Catalog No. 50-004-11769 Supplier Medchemexpress LLC Supplier No. HYB1057100MG
Nefopam hydrochloride is a centrally-acting, non-opioid, and non-steroidal analgesic drug. It is derived from benzoxazocine and is used for the relief of moderate to severe pain. It targets β-catenin protein levels in mesenchymal cells both in-vitro and in-vivo, inhibiting β-catenin mediated signaling during skin wound repair.
  • Centrally-acting analgesic
  • Non-opioid and non-steroidal properties
  • Aids in the relief of moderate to severe pain
  • Inhibits β-catenin mediated signaling
  • Derived from benzoxazocine