Learn More
Invivogen Negative control for ODN 2216, 1 mg
SDP
Catalog No. NC0731756
Encompass
ODN 2243 (also known as ODN 2216 Control) is designed as a negative control for the TLR9 agonist ODN 2216.
Catalog No. NC0731756 Supplier Invivogen Supplier No. TLRL22431
Add to Cart
Add to Cart
ODN 2243 (also known as ODN 2216 Control) is designed as a negative control for the TLR9 agonist ODN 2216.