Learn More
AllCells LLC Peripheral Blood, Cryopreserved, CD14+ Monocytes, Negatively Selected, 10M
Catalog No. 50156878
Encompass
AllCells Untouched NPB-CD14+ Monocytes are indirectly isolated from mononuclear cells using an immunomagnetic labeling system.
Catalog No. 50-156-878 Supplier AllCells LLC Supplier No. LP, CR, CD14+, NS, 10M
AllCells Untouched NPB-CD14+ Monocytes are indirectly isolated from mononuclear cells using an immunomagnetic labeling system.