Learn More
AllCells LLC Peripheral Blood, Cryopreserved, CD56+ Natural Killer (NK) Cells, Negatively Selected, 5M
Catalog No. 50156887
Encompass
CD56+ Natural Killer (NK) cells are negatively selected from peripheral blood leukapheresis using immunomagnetic microbeads targeting non-NK cells. CD56+ NK cells are shipped in a 2mL cryovial. Purity is determined prior to cryopreservation and is ≥ 85%. Viability is ≥ 80% post-thaw.
Catalog No. 50-156-887 Supplier AllCells LLC Supplier No. LP, CR, CD56+, NS, 5M
CD56+ Natural Killer (NK) cells are negatively selected from peripheral blood leukapheresis using immunomagnetic microbeads targeting non-NK cells. CD56+ NK cells are shipped in a 2mL cryovial. Purity is determined prior to cryopreservation and is ≥ 85%. Viability is ≥ 80% post-thaw.