Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Negative Cellularity Control (Inactivated Swab)
SDP

Catalog No. 23047382 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-047-382 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-047-382 Supplier Microbiologics™ Supplier No. HE0067NS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inactivated Helix Elite Molecular Standards are specially designed, full process controls for use in molecular testing

  • Inactivated Helix Elite Molecular Standards are specially designed, full process controls for use in molecular testing.
  • Each kit consists of 6 individually packaged lyophilized swabs that contain human lung epithelial cells.
  • Each kit contains six positive and six negative, inactivated control swabs (12 swabs/kit).
  • Positive Control (six swabs): Influenza A (H3N2) Virus, Influenza A (H1N1) Virus, Influenza B Virus, Influenza B Virus, Respiratory Syncytial Virus, SARS-CoV-2 Virus, Human lung epithelial cells.
  • Negative Control (six swabs): Coxsackievirus B1, Human lung epithelial cells

Specifications

Biosafety Level 1
Content And Storage 2 to 25°C
Format Swab
Manufacturing Origin United States
Packaging Type Plastic Box
Quantity 6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.