Learn More
Invivogen ODN 1826 Control (ODN 2138), Synthetic Oligonucleotide, Nuclease Resistant, 1 mg
SDP
Catalog No. NC3253202
Encompass
The ODN 1826 Control (ODN 2138) is a synthetic oligonucleotide designed as a negative control for experiments involving ODN 1826, specific for murine TLR9. It features GpC dinucleotides to ensure observed effects are due to CpG recognition. Enhanced nuclease resistance and rigorous testing for contamination make it a reliable choice for researchers needing a controlled environment for TLR9-targeted studies.
Catalog No. NC3253202 Supplier Invivogen Supplier No. TLRL1826C1
The ODN 1826 Control (ODN 2138) is a synthetic oligonucleotide designed as a negative control for experiments involving ODN 1826, specific for murine TLR9. It features GpC dinucleotides to ensure observed effects are due to CpG recognition. Enhanced nuclease resistance and rigorous testing for contamination make it a reliable choice for researchers needing a controlled environment for TLR9-targeted studies.