Learn More
Enzo Life Sciences RGD control peptide (5 mg)
SDP
Supplier:  Enzo Life Sciences BMLP7010005
Encompass_Preferred
Negative control for RGD peptide. Inactive control peptide. Alternative Name: GRADSP. MW: 601.6. Formulation: Lyophilized. Purity: ≥97% (HPLC). Sequence Gly-Arg-Ala-Asp-Ser-Pro. Appearance: White to off-white powder. Solubility: Soluble in Water. Long Term Storage: -20°C
Catalog No. NC1225518