Learn More
Biogenex Laboratories Negative Control Sera/Immunoglobulins
SDP
Catalog No. NC1439462
Encompass_Preferred
Since each staining run should include a negative control slide to confirm reagent specificity, we offer negative control sera/immunoglobulins which have been optimized for use as negative controls for our Super Sensitive detection systems and ready-to-use antibodies. These controls are available for mouse, goat, rat and rabbit antibodies.
Catalog No. NC1439462 Supplier Biogenex Laboratories Supplier No. HK1197M
Since each staining run should include a negative control slide to confirm reagent specificity, we offer negative control sera/immunoglobulins which have been optimized for use as negative controls for our Super Sensitive detection systems and ready-to-use antibodies. These controls are available for mouse, goat, rat and rabbit antibodies.