Learn More
Biotium Negative Control for Rabbit Monoclonal Antibodies (NCRBM/1520R), CF640R conjugate, 0.1mg/mL
Catalog No. 5000657656
Encompass_Preferred
To serve as a true negative control for various procedures, this rabbit monoclonal IgG should be used under the identical conditions and at the same dilution as the rabbit monoclonal antibody being used for a positive reaction. Available purified, or with a selection of fluorescent CF Dyes and other labels. CF Dyes offer exceptional brightness and photostability.
Catalog No. 50-006-57656 Supplier Biotium Supplier No. BNC401520500
Add to Cart
Add to Cart
To serve as a true negative control for various procedures, this rabbit monoclonal IgG should be used under the identical conditions and at the same dilution as the rabbit monoclonal antibody being used for a positive reaction. Available purified, or with a selection of fluorescent CF Dyes and other labels. CF Dyes offer exceptional brightness and photostability.