Learn More
Aviva Systems Biology Corp NEGR1 Antibody - N-terminal region (ARP55701_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55701_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NEGR1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human NEGR1. Alternate names: DMML2433, IGLON4, KILON, MGC46680, NtraApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-066