Learn More
Biomatik Corporation Negr1 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11888
Encompass_Preferred
Negr1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NEGR1. The Negr1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Neuronal Growth Regulator 1 Protein (1-196Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: NEGR1. Accession Number: Q7Z3B1. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6502