Learn More
Bioss Neisseria gonorrhoeae
SDP
Catalog No. 5000066117
Encompass_Preferred
Neisseria gonorrhoeae Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-66117 Supplier Bioss Supplier No. BSM72340M
Neisseria gonorrhoeae Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide