Learn More
ATCC Neisseria gonorrhoeae
Supplier:  ATCC 49226
Encompass
Neisseria gonorrhoeae strain F-18 [89-018314, CDC 10,001, P935] is a whole-genome sequenced bacterium that can be used for susceptibility disc testing, media testing, and sexually transmitted disease research.
Catalog No. 50-238-2443