Learn More
ATCC Neisseria gonorrhoeae
Supplier:  ATCC 19424
Encompass
Neisseria gonorrhoeae strain NCTC 8375 [B 5025] is a whole-genome sequenced bacterial type strain that has applications in quality control, media testing, and infectious disease research.
Catalog No. 50-238-4817