Learn More
ATCC Streptococcus; gallolyticus; subsp. gallolyticus
Catalog No. 502381195
Encompass
Streptococcus gallolyticus subsp. gallolyticus strain AmMS 236 is a bacterial clinical isolate. This culture can be used as a quality control strain.
Catalog No. 50-238-1195 Supplier ATCC Supplier No. 49475
Streptococcus gallolyticus subsp. gallolyticus strain AmMS 236 is a bacterial clinical isolate. This culture can be used as a quality control strain.