Learn More
ATCC Pfizer Pneumoccal polysaccharide powder Type 70 (d33F) 2mg
Catalog No. 502381147
Encompass
Purified pneumococcal polysaccharide type 33F is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 33F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-1147 Supplier ATCC Supplier No. 533X
Purified pneumococcal polysaccharide type 33F is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 33F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.