Learn More
ATCC Quantitative Genomic DNA from Neisseria gonorrhoeae strain FA1090 (ATCC 700825DQ™)
Catalog No. 502382808
Encompass
Quantitative Genomic DNA from Neisseria gonorrhoeae strain FA1090 (ATCC 700825DQ™) is a molecular standard used for qPCR assay development, validation, and bacterial load quantification. Suitable for sexually transmitted infection research and bioinformatics applications.
Catalog No. 50-238-2808 Supplier ATCC Supplier No. 700825DQ
Quantitative Genomic DNA from Neisseria gonorrhoeae strain FA1090 (ATCC 700825DQ™) is a molecular standard used for qPCR assay development, validation, and bacterial load quantification. Suitable for sexually transmitted infection research and bioinformatics applications.