Learn More
Creative Biomart Recombinant Lassa Nucleoprotein Antigen, His-tagged
Supplier:  Creative Biomart VAC0337
Encompass_Preferred
The E. coli derived Recombinant Lassa Nucleoprotein (LASV strain AV, GenBank accession number AAG41803) containing 420 amino acids of Lassa virus nucleoprotein.The Lassa Capsid protein is fused to a 6
Catalog No. 50-208-5177