Learn More
Creative Biomart Recombinant HIV-1 Subtype B gp41 Antigen, His-tagged
Supplier:  Creative Biomart VAC0334
Encompass_Preferred
Recombinant HIV-1 gp41 Subtype B produced in E. coli is a non-glycosylated polypeptide chain having a molecular mass of 16 kDa and fused to a His tag at N-terminus.
Catalog No. 50-208-5174