Learn More
Creative Biomart Recombinant HCV Core nucleocapsid Antigen, HRP-Labeled
Supplier:  Creative Biomart VAC0332
Encompass_Preferred
The E. coli derived recombinant HRP Labeled protein contains the HCV core nucleocapsid immunodominant regions, amino acids 2-119.
Catalog No. 50-208-5172