Learn More
Creative Biomart Recombinant HIV-1 gag p17-p24 and gp41-gp120 Antigen
Supplier:  Creative Biomart VAC0333
Encompass_Preferred
HIV-1 p17-p24, gp-41-gp120 is a non-glycosylated polypeptide chain, containing sequence of HIV-1 immunodominant regions p17-p24, gp41-gp120.
Catalog No. 50-208-5173