Learn More
Creative Biomart Recombinant Feline Leukemia Virus p27 Antigen, His-tagged
Supplier:  Creative Biomart VAC0330
Encompass_Preferred
The E. coli truncated derived recombinant protein contains the p27 immunodominant regions, containing 248 amino acids (271-519) and having a Mw of 31 kDa as visualized by SDS-PAGE. Recombinant Feline
Catalog No. 50-208-5170
 Restricted Item