Learn More
Creative Biomart Recombinant Streptococcus pneumoniae type 3 Capsular Polysaccharide Antigen
Supplier:  Creative Biomart VAC0259
Encompass_Preferred
Streptococcus pneumoniae type 3 capsular polysaccharide antigen is a vaccine that contains highly purified capsular polysaccharides from the invasive pneumococcal type 3 of Streptococcus pneumoniae. I
Catalog No. 50-208-5099