Learn More
Creative Biomart Recombinant HBsAg adw2 Antigen, His-tagged
Supplier:  Creative Biomart VAC0331
Encompass_Preferred
HbsAg subtype adw2 produced in E. coli, is a single, non-glycosylated, polypeptide chain containing 226 amino acids and having a molecular weight of approximately 23.0kDa. HBsAg adw2 migrates on SDS-P
Catalog No. 50-208-5171