Learn More
ATCC Neisseria sicca
Supplier:  ATCC 9913
Encompass
Produces restriction endonuclease NsiAI. Used in quality assurance for commercially prepared microbiological culture media.
Catalog No. 50-238-3782