Learn More
Carnabio Usa Inc NEK2/10UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 0522610
Encompass
NEK2, Full-length, Wild type, Amino Acid 1-445, NP_002488.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0897