Learn More
Sino Biological Recombinant Rat NEK7 Protein (full length, GST Tag)
Catalog No. 5000083638
Encompass_Preferred
Recombinant full-length rat NEK7 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-000-83638 Supplier Sino Biological Supplier No. N09A10G10
Recombinant full-length rat NEK7 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.