Learn More
Sino Biological Recombinant Rabbit NEK7 Protein (full length, GST Tag)
Catalog No. 5000084307
Encompass_Preferred
Recombinant full-length rabbit NEK7 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-000-84307 Supplier Sino Biological Supplier No. N09R10G10
Add to Cart
Add to Cart
Recombinant full-length rabbit NEK7 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.