Learn More
Sino Biological Recombinant Rabbit NEK7 Protein (full length, GST Tag)
Catalog No. 5000083692
Encompass_Preferred
Recombinant full-length rabbit NEK7 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-000-83692 Supplier Sino Biological Supplier No. N09R10G50
Recombinant full-length rabbit NEK7 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.