Learn More
Sino Biological Recombinant Cynomolgus NEK7 Protein (full length, GST Tag)
Catalog No. 5000083874
Encompass_Preferred
Recombinant full-length monkey NEK7 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-000-83874 Supplier Sino Biological Supplier No. N09C10G50
Recombinant full-length monkey NEK7 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.