Learn More
Aviva Systems Biology Corp NEK7 Antibody - N-terminal region (ARP48999_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48999_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NEK7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NEK7. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-59989