Learn More
Medchemexpress LLC Nek7 protein, human (His-SUMO) | >90.0% | 20UG

Supplier: Medchemexpress LLC HYP70125320UG
Recombinant full-length human NEK7 protein, N-terminally tagged with 6xHis and SUMO, expressed in E. coli and supplied lyophilized for biochemical and structural studies. The protein is provided as a purified preparation (>90% by reducing SDS-PAGE), with an approximate molecular weight of 50 kDa and sequence corresponding to isoform 1 (M1-S302). Lyophilized from PBS with 6% trehalose; store at -20°C. After reconstitution, stable at 4°C for 1 week or -20°C for longer with carrier protein.
- Full-length human NEK7 kinase (M1-S302).
- N-terminal 6xHis and SUMO tags for purification.
- Expressed in E. coli for high-yield production.
- Purity >90% as determined by reducing SDS-PAGE.
- Lyophilized formulation in PBS with 6% trehalose.
- Stable storage at -20°C; short-term stability at 4°C after reconstitution.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.