Learn More
Eurofins Discoverx NEK9, active, 10 µg. (14-936)
Supplier:  Eurofins Discoverx 14936
Encompass
N-terminal 6xHis-tagged recombinant, human NEK9 amino acids 1-324. Pleiotropic regulator of mitotic progression, participating in the control of spindle dynamics and chromosome separation. Phosphorylates different histones, myelin basic protein, beta-casein, and BICD2. Phosphorylates histone H3 on serine and threonine amino acids and beta-casein on serine residues. Important for G1/S transition and S phase progression. Phosphorylates NEK6 and NEK7 and stimulates their activity by releasing the autoinhibitory functions of Tyr-108 and Tyr-97 respectively. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9445