Learn More
TARGETMOL CHEMICALS INC NELARABINE 10MG
SDP
Catalog No. 502579327
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Nelarabine (GW 506U78) is a purine nucleoside analog and DNA synthesis inhibitor with IC50 from 0.067-2.15 uM in tumor cells. purity: 99%
Catalog No. 50-257-9327 Supplier TARGETMOL CHEMICALS INC Supplier No. T660310MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Nelarabine (GW 506U78) is a purine nucleoside analog and DNA synthesis inhibitor with IC50 from 0.067-2.15 uM in tumor cells. purity: 99%