Learn More
Aviva Systems Biology Corp NELF Antibody - N-terminal region (ARP42448_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42448_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NELF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NELF. Alternate names: MGC125369, RP11-48C7.1, NELFApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-58228