Learn More
Medchemexpress LLC Nelfinavir | 159989-64-7 | 98.65% | 567.78 | 50 MG
SDP
Catalog No. 5000364112
Encompass_Preferred
Nelfinavir is a potent, orally bioavailable HIV-1 protease inhibitor and a broad-spectrum anticancer agent. It is intended for research use only.
  • Potent and orally bioavailable HIV-1 protease inhibitor
  • Broad-spectrum anticancer agent
  • Inhibits proliferation of multiple myeloma cells
  • Induces apoptosis
  • Decreases phosphorylation of AKT
Catalog No. 50-003-64112 Supplier Medchemexpress LLC Supplier No. 50MGHY15287
Nelfinavir is a potent, orally bioavailable HIV-1 protease inhibitor and a broad-spectrum anticancer agent. It is intended for research use only.
  • Potent and orally bioavailable HIV-1 protease inhibitor
  • Broad-spectrum anticancer agent
  • Inhibits proliferation of multiple myeloma cells
  • Induces apoptosis
  • Decreases phosphorylation of AKT