Learn More
Medchemexpress LLC Nelfinavir | 159989-64-7 | C32H45N3O4S | 1 MG
SDP
Catalog No. 5000372033
Encompass_Preferred
Nelfinavir is a potent, orally bioavailable HIV-1 protease inhibitor, showing strong efficacy against HIV infection with a Ki of 2 nM. This compound also functions as a broad-spectrum anticancer agent.
  • Potent and orally bioavailable HIV-1 protease inhibitor
  • Broad-spectrum anticancer agent
  • Inhibits proliferation of multiple myeloma cells
  • Inhibits 26S chymotrypsin-like proteasome activity
  • Triggers apoptosis in myeloma cell lines and fresh plasma cells
  • Decreases phosphorylation of AKT, STAT-3, ERK1/2
  • Activates pro-apoptotic pathway of the unfolded protein response system
  • Decreases multiple myeloma cell growth in NOD/SCID mice
Catalog No. 50-003-72033 Supplier Medchemexpress LLC Supplier No. HY152871MG
Nelfinavir is a potent, orally bioavailable HIV-1 protease inhibitor, showing strong efficacy against HIV infection with a Ki of 2 nM. This compound also functions as a broad-spectrum anticancer agent.
  • Potent and orally bioavailable HIV-1 protease inhibitor
  • Broad-spectrum anticancer agent
  • Inhibits proliferation of multiple myeloma cells
  • Inhibits 26S chymotrypsin-like proteasome activity
  • Triggers apoptosis in myeloma cell lines and fresh plasma cells
  • Decreases phosphorylation of AKT, STAT-3, ERK1/2
  • Activates pro-apoptotic pathway of the unfolded protein response system
  • Decreases multiple myeloma cell growth in NOD/SCID mice