Learn More
Medchemexpress LLC Nelfinavir | 159989-64-7 | C32H45N3O4S | 1 ML
SDP
Catalog No. 5000372131
Encompass_Preferred
Nelfinavir is a potent, orally bioavailable HIV-1 protease inhibitor with a Ki of 2 nM for HIV infection. It is also recognized as a broad-spectrum anticancer agent. This product is supplied as a 10 mM solution in DMSO.
  • Potent HIV-1 protease inhibitor (Ki=2 nM)
  • Orally bioavailable
  • Broad-spectrum anticancer agent
  • For research use only
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-72131 Supplier Medchemexpress LLC Supplier No. HY1528710MM/1ML
Add to Cart
Add to Cart
Nelfinavir is a potent, orally bioavailable HIV-1 protease inhibitor with a Ki of 2 nM for HIV infection. It is also recognized as a broad-spectrum anticancer agent. This product is supplied as a 10 mM solution in DMSO.
  • Potent HIV-1 protease inhibitor (Ki=2 nM)
  • Orally bioavailable
  • Broad-spectrum anticancer agent
  • For research use only
  • Store in solvent at -80°C for 6 months or -20°C for 1 month