Learn More
Medchemexpress LLC Nelfinavir | 159989-64-7 | MFCD01938163 | 98.8% | 567.78 g·mol⁻¹ | C32H45N3O4S | 10 MG
SDP
Catalog No. 5000285735
Encompass_Preferred
Nelfinavir is a research-grade small-molecule inhibitor of HIV-1 protease (Ki ≈ 2 nM) supplied as a powder for laboratory research and analytical applications. It has also been reported to show broad-spectrum anticancer activity in preclinical studies. Intended for research use only; not for human or veterinary use.
  • Potent HIV-1 protease inhibitor with low nanomolar affinity.
  • Reported anticancer activity in preclinical models.
  • Supplied as a powder for convenient handling and storage.
  • Available in small pack sizes suitable for screening and assay development.
  • Stable under recommended storage conditions (powder: -20°C/3 years; 4°C/2 years).
Catalog No. 50-002-85735 Supplier Medchemexpress LLC Supplier No. HY1528710MG
Nelfinavir is a research-grade small-molecule inhibitor of HIV-1 protease (Ki ≈ 2 nM) supplied as a powder for laboratory research and analytical applications. It has also been reported to show broad-spectrum anticancer activity in preclinical studies. Intended for research use only; not for human or veterinary use.
  • Potent HIV-1 protease inhibitor with low nanomolar affinity.
  • Reported anticancer activity in preclinical models.
  • Supplied as a powder for convenient handling and storage.
  • Available in small pack sizes suitable for screening and assay development.
  • Stable under recommended storage conditions (powder: -20°C/3 years; 4°C/2 years).