Learn More
Medchemexpress LLC HY-15287A 10mg , Nelfinavir (Mesylate) CAS:159989-65-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15287A10MG
Encompass_Preferred
HY-15287A 10mg Nelfinavir (Mesylate) CAS: 159989-65-8 Nelfinavir Mesylate (AG 1343 Mesylate) is a potent and orally bioavailable HIV-1 protease inhibitor (Ki=2 nM) for HIV infection. Nelfinavir Mesylate (AG 1343 Mesylate) is a broad-spectrum, anticancer agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0156