Learn More
Apexbio Technology LLC Nelfinavir Mesylate 159989-65-8 5mg
SDP
Catalog No. 5000093705
Encompass_Preferred
Nelfinavir Mesylate (CAS 159989-65-8) is a small-molecule inhibitor targeting HIV-1 protease It is designed to inhibit the enzymatic processing of gag and gag-pol precursor polyproteins thereby interfering with viral maturation Nelfinavir Mesylate exerts its biological activity primarily through inhibition of HIV-1 protease activity In cell-based antiviral assays using HIV-1-infected cells Nelfinavir Mesylate demonstrates inhibition of viral replication with EC50 values ranging from 14 nM to 43 nM Based on these pharmacological properties Nelfinavir Mesylate holds research potential in preclinical studies investigating HIV replication mechanisms therapeutic resistance profiles and combination antiretroviral drug strategies
Catalog No. 50-000-93705 Supplier Apexbio Technology LLC Supplier No. A36535
Add to Cart
Add to Cart
Nelfinavir Mesylate (CAS 159989-65-8) is a small-molecule inhibitor targeting HIV-1 protease It is designed to inhibit the enzymatic processing of gag and gag-pol precursor polyproteins thereby interfering with viral maturation Nelfinavir Mesylate exerts its biological activity primarily through inhibition of HIV-1 protease activity In cell-based antiviral assays using HIV-1-infected cells Nelfinavir Mesylate demonstrates inhibition of viral replication with EC50 values ranging from 14 nM to 43 nM Based on these pharmacological properties Nelfinavir Mesylate holds research potential in preclinical studies investigating HIV replication mechanisms therapeutic resistance profiles and combination antiretroviral drug strategies