Learn More
Medchemexpress LLC Nelfinavir mesylate | 159989-65-8 | C33H49N3O7S2 | 1 ML
SDP
Catalog No. 5000372115
Encompass_Preferred
Nelfinavir mesylate (AG 1343 Mesylate) is a potent and orally bioavailable HIV-1 protease inhibitor (Ki=2 nM) used for HIV infection. It also acts as a broad-spectrum anticancer agent.
  • Potent HIV-1 protease inhibitor (Ki=2 nM)
  • Broad-spectrum anticancer agent
  • Inhibits proliferation of multiple myeloma cells
  • Triggers apoptosis in myeloma cell lines
  • Decreases phosphorylation of AKT
  • Activates cleavage of caspase-3
  • SARS-CoV 3CLpro inhibitor (IC50 = 35.93 μM)
Catalog No. 50-003-72115 Supplier Medchemexpress LLC Supplier No. HY15287A10MM/1ML
Nelfinavir mesylate (AG 1343 Mesylate) is a potent and orally bioavailable HIV-1 protease inhibitor (Ki=2 nM) used for HIV infection. It also acts as a broad-spectrum anticancer agent.
  • Potent HIV-1 protease inhibitor (Ki=2 nM)
  • Broad-spectrum anticancer agent
  • Inhibits proliferation of multiple myeloma cells
  • Triggers apoptosis in myeloma cell lines
  • Decreases phosphorylation of AKT
  • Activates cleavage of caspase-3
  • SARS-CoV 3CLpro inhibitor (IC50 = 35.93 μM)