Learn More
Medchemexpress LLC Nelfinavir mesylate | 159989-65-8 | C33H49N3O7S2 | 25 MG
SDP
Catalog No. 5000374726
Encompass_Preferred
Nelfinavir Mesylate, also known as AG 1343 Mesylate, is a potent and orally bioavailable HIV-1 protease inhibitor. It is also described as a broad-spectrum anticancer agent and an inhibitor of SARS-CoV 3CLpro.
  • Potent HIV-1 protease inhibitor (Ki=2 nM)
  • Inhibits proliferation of multiple myeloma cells (IC50 of 1-5 μM)
  • Triggers apoptosis of myeloma cell lines
  • Decreases phosphorylation of AKT, STAT-3, ERK1/2
  • Activates the pro-apoptotic pathway of the unfolded protein response system
  • Inhibits SARS-CoV 3CLpro (IC50 of 35.93 μM)
  • Decreases multiple myeloma cell growth in animal models
Catalog No. 50-003-74726 Supplier Medchemexpress LLC Supplier No. 25MGHY15287A
Nelfinavir Mesylate, also known as AG 1343 Mesylate, is a potent and orally bioavailable HIV-1 protease inhibitor. It is also described as a broad-spectrum anticancer agent and an inhibitor of SARS-CoV 3CLpro.
  • Potent HIV-1 protease inhibitor (Ki=2 nM)
  • Inhibits proliferation of multiple myeloma cells (IC50 of 1-5 μM)
  • Triggers apoptosis of myeloma cell lines
  • Decreases phosphorylation of AKT, STAT-3, ERK1/2
  • Activates the pro-apoptotic pathway of the unfolded protein response system
  • Inhibits SARS-CoV 3CLpro (IC50 of 35.93 μM)
  • Decreases multiple myeloma cell growth in animal models