Learn More
Selleck Chemical LLC Nelfinavir Mesylate S4282-10mM/1mL
SDP
Catalog No. 503110175
Encompass_Preferred
Nelfinavir Mesylate is a potent HIV protease inhibitor with Ki of 2 nM
Catalog No. 50-311-0175 Supplier Selleck Chemical LLC Supplier No. S428210MM/1ML
Nelfinavir Mesylate is a potent HIV protease inhibitor with Ki of 2 nM