Learn More
TARGETMOL CHEMICALS INC NELFINAVIR MESYLATE 25MG
SDP
Catalog No. 502579704
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Nelfinavir Mesylate (Viracept) is an effective HIV protease inhibitor (Ki 2 nM). purity: 98%
Catalog No. 50-257-9704 Supplier TARGETMOL CHEMICALS INC Supplier No. T6001
Add to Cart
Add to Cart
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Nelfinavir Mesylate (Viracept) is an effective HIV protease inhibitor (Ki 2 nM). purity: 98%