Learn More
Sino Biological NELL2 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 13132MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human NELL2 (rh NELL2; Catalog#13132-H08B; Q99435; Met1-Leu816). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8692