Learn More
Sino Biological NELL2 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 13132R055100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human NELL2 (rh NELL2; Catalog#13132-H08B; Q99435; Met1-Leu816).
Catalog No. 50-224-7916