Learn More
Sino Biological NELL2 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 13132T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human NELL2 (rh NELL2; Catalog#13132-H08B; Q99435; Met1-Leu816). NELL2 specific IgG was purified by Human NELL2 affinity chromatography.
Catalog No. 50-224-8696