Learn More
Medchemexpress LLC Nelociguat (BAY60-4552) | 625115-52-8 | 99.7% | C19H17FN8O2 | 100 MG
SDP
Catalog No. 5000433158
Encompass_Preferred
Nelociguat (BAY60-4552) is a nitric oxide sensitive soluble guanylate cyclase stimulator. It is involved in the nitric oxide signalling pathway and has shown potential benefits in hypertension-induced end-organ damage and erectile dysfunction.
  • Functions as a nitric oxide sensitive soluble guanylate cyclase stimulator.
  • Involved in the nitric oxide signalling pathway.
  • May provide partial benefit against hypertension-induced end-organ damage.
  • Decreases urine output and improves survival in hypertensive rats.
  • Reduces microalbuminuria and attenuates increased mean arterial pressure.
  • Shows synergistic beneficial effects for erectile dysfunction when combined with vardenafil.
Catalog No. 50-004-33158 Supplier Medchemexpress LLC Supplier No. HY78237100MG
Nelociguat (BAY60-4552) is a nitric oxide sensitive soluble guanylate cyclase stimulator. It is involved in the nitric oxide signalling pathway and has shown potential benefits in hypertension-induced end-organ damage and erectile dysfunction.
  • Functions as a nitric oxide sensitive soluble guanylate cyclase stimulator.
  • Involved in the nitric oxide signalling pathway.
  • May provide partial benefit against hypertension-induced end-organ damage.
  • Decreases urine output and improves survival in hypertensive rats.
  • Reduces microalbuminuria and attenuates increased mean arterial pressure.
  • Shows synergistic beneficial effects for erectile dysfunction when combined with vardenafil.